Learn More
American Research Products Inc Rat Insulin like growth factor
Supplier:  American Research Products Inc CSBE07353R
Encompass
Rat Insulin-like growth factor binding protein 3 IGFBP-3 ELISA kit from CUSABIO Detection Range 0 24 ng/mL-15 ng/mL Sensitivity 0 06 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26467