Learn More
American Research Products Inc Rat Insulin like growth factor
Supplier:  American Research Products Inc CSBE13873R
Encompass
Rat Insulin-like growth factor 1 receptor IGF-1R ELISA Kit from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 8 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28318