Learn More
Biomatik Corporation Rat Insulin Like Growth Factor 2 (IGF2) ELISA Kit
Supplier:  Biomatik Corporation EKU04979
Encompass_Preferred
Rat Insulin Like Growth Factor 2 (IGF2) ELISA Kit for detecting IGF2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78-5,000pg/ml. Sensitivity: 33pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: p01346. Gene ID: 24483. Target Synonyms: INSIGF; IGF-II; Somatomedin A; Preptin; Insulin-like growth factor II Ala-25 Del. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5972