Learn More
Biomatik Corporation Rat Insulin Like Protein 3 (INSL3) ELISA Kit
Supplier:  Biomatik Corporation EKU12028
Encompass_Preferred
Rat Insulin Like Protein 3 (INSL3) ELISA Kit for detecting INSL3 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.12-200pg/ml. Sensitivity: 1.17pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9WUK0. Gene ID: 114215. Target Synonyms: RLNL; RLF; Relaxin-Like Factor,Leydig Cell; Leydig insulin-like peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8049