Learn More
Biomatik Corporation Rat Insulin Like Protein 3 (INSL3) ELISA Kit
Supplier:  Biomatik Corporation EKU05014
Encompass_Preferred
Rat Insulin Like Protein 3 (INSL3) ELISA Kit for detecting INSL3 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 185.19-15000pg/ml. Sensitivity: 62.8pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q9WUK0. Gene ID: 114215. Target Synonyms: RLNL; RLF; Relaxin-Like Factor,Leydig Cell; Leydig insulin-like peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5700