Learn More
Biomatik Corporation Rat Insulin Receptor Substrate 3 (IRS3) ELISA Kit
Supplier:  Biomatik Corporation EKU05025
Encompass_Preferred
Rat Insulin Receptor Substrate 3 (IRS3) ELISA Kit for detecting IRS3 in tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.227ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: B1WBQ1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6248