Learn More
Quidel Corporation MicroVue Rat Intact PTH ELISA Kit
SDP
Supplier:  Quidel Corporation 602500
Encompass
The MicroVue Rat Intact PTH ELISA Kit is for the measurement of PTH in rat serum, plasma or cell culture media.
Catalog No. NC9428444