Learn More
Biomatik Corporation Rat Inter Alpha-Globulin Inhibitor H2 (ITIH2) ELISA Kit
Supplier:  Biomatik Corporation EKU11801
Encompass_Preferred
Rat Inter Alpha-Globulin Inhibitor H2 (ITIH2) ELISA Kit for detecting ITIH2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: A0A8I5ZVC4. Target Synonyms: H2P; SHAP; IGHEP2; Serum-derived hyaluronan-associated protein; Inter-alpha-trypsin inhibitor complex component II. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8079