Learn More
American Research Products Inc Rat intercellular adhesion mol
Catalog No. 5000328762
Encompass
Rat intercellular adhesion molecule 1 ICAM-1 ELISA Kit from CUSABIO Detection Range 78 125 pg/mL-5000 pg/mL Sensitivity 24 321 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-28762 Supplier American Research Products Inc Supplier No. CSBE04576R
Rat intercellular adhesion molecule 1 ICAM-1 ELISA Kit from CUSABIO Detection Range 78 125 pg/mL-5000 pg/mL Sensitivity 24 321 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich