Learn More
Biomatik Corporation Rat Interferon Alpha 13 (IFNa13) ELISA Kit
Supplier:  Biomatik Corporation EKU05082
Encompass_Preferred
Rat Interferon Alpha 13 (IFNa13) ELISA Kit for detecting IFNa13 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 7.81-500pg/ml. Sensitivity: 2.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P05011. Gene ID: 298210. Target Synonyms: LeIF D; Interferon alpha-D. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6364