Learn More
American Research Products Inc Rat Interferon b IFN b IFNB E
Catalog No. 5000326522
Encompass
Rat Interferon b IFN-b/IFNB ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26522 Supplier American Research Products Inc Supplier No. CSBE04845R
Rat Interferon b IFN-b/IFNB ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich