Learn More
American Research Products Inc Rat interferon inducible prote
Catalog No. 5000326150
Encompass
Rat interferon-inducible protein 10 IP-10 ELISA Kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 31 25 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26150 Supplier American Research Products Inc Supplier No. CSBE08182RIS
Add to Cart
Add to Cart
Rat interferon-inducible protein 10 IP-10 ELISA Kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 31 25 pg/mL Sample serum plasma tissue homogenates Method Sandwich