Learn More
American Research Products Inc Rat interferon inducible prote
Catalog No. 5000326440
Encompass
Rat interferon-inducible protein 10 IP-10 ELISA Kit from CUSABIO Detection Range 781 25 pg/mL-50000 pg/mL Sensitivity 195 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26440 Supplier American Research Products Inc Supplier No. CSBE08182R
Rat interferon-inducible protein 10 IP-10 ELISA Kit from CUSABIO Detection Range 781 25 pg/mL-50000 pg/mL Sensitivity 195 pg/mL Sample serum plasma tissue homogenates Method Sandwich