Learn More
Biomatik Corporation Rat Interleukin 1 Receptor Type I (IL1R1) ELISA Kit
Supplier:  Biomatik Corporation EKU12113
Encompass_Preferred
Rat Interleukin 1 Receptor Type I (IL1R1) ELISA Kit for detecting IL1R1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.060ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q02955. Target Synonyms: CD121a; CD121-A; IL1-R1; IL1R; IL1RA; P80; CD121 Antigen-Like Family Member A; Interleukin-1 Receptor Alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7782