Learn More
American Research Products Inc Rat Interleukin 12 IL 12 P40
Catalog No. 5000326470
Encompass
Rat Interleukin 12 IL-12/P40 ELISA KIT from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26470 Supplier American Research Products Inc Supplier No. CSBE07362R
Rat Interleukin 12 IL-12/P40 ELISA KIT from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma tissue homogenates Method Sandwich