Learn More
American Research Products Inc Rat Interleukin 12 IL 12 P70
Catalog No. 5000328957
Encompass
Rat Interleukin 12 IL-12/P70 ELISA KIT from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28957 Supplier American Research Products Inc Supplier No. CSBE07364R
Rat Interleukin 12 IL-12/P70 ELISA KIT from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma tissue homogenates Method Sandwich