Learn More
American Research Products Inc Rat Interleukin 17 IL 17 ELIS
Catalog No. 5000328859
Encompass
Rat Interleukin 17 IL-17 ELISA KIT from CUSABIO Detection Range 93 75 pg/mL-6000 pg/mL Sensitivity 23 43 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28859 Supplier American Research Products Inc Supplier No. CSBE07451R
Rat Interleukin 17 IL-17 ELISA KIT from CUSABIO Detection Range 93 75 pg/mL-6000 pg/mL Sensitivity 23 43 pg/mL Sample serum plasma tissue homogenates Method Sandwich