Learn More
Biomatik Corporation Rat Interleukin 17B (IL17B) CLIA Kit
Supplier:  Biomatik Corporation EKU09699
Encompass_Preferred
Rat Interleukin 17B (IL17B) CLIA Kit for detecting IL17B in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.51pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: G3V8K9. Gene ID: 116472. Target Synonyms: IL-20; Cytokine Zcyto7; Neuronal interleukin-17-related factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8462