Learn More
Biomatik Corporation Rat Interleukin 17F (IL17F) CLIA Kit
Supplier:  Biomatik Corporation EKU09809
Encompass_Preferred
Rat Interleukin 17F (IL17F) CLIA Kit for detecting IL17F in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.64pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q5BJ95. Gene ID: 301291. Target Synonyms: ML-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6731