Learn More
Biomatik Corporation Rat Interleukin 18 (IL18) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11178
Encompass_Preferred
Rat Interleukin 18 (IL18) Mini sample ELISA Kit for detecting IL18 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P97636. Gene ID: 29197. Target Synonyms: IGIF; IL-1g; IL1F4; IL1-F4; Interferon-Gamma-Inducing Factor; Interleukin-1 Family Member 4; Iboctadekin; Interleukin-1 Gamma. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7288