Learn More
Bioworld RAT INTERLEUKIN-2 (RECOMBIN
Supplier:  Bioworld 220606661
Encompass_Preferred
RAT INTERLEUKIN-2 (RECOMBINANT): Recombinant Rat Interleukin-2 (IL-2) Source: E.Coli Description: Recombinant Rat IL-2 produced in E.Coli is a single, non-glycosylated polypeptide chain containing 134 amino acids and having a molecul...
Catalog No. 50-253-762