Learn More
American Research Products Inc Rat Interleukin 22 IL 22 ELI
Catalog No. 5000328495
Encompass
Rat Interleukin 22 (IL-22) ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28495 Supplier American Research Products Inc Supplier No. CSBE14918R
Rat Interleukin 22 (IL-22) ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich