Learn More
Biomatik Corporation Rat Interleukin 23 Receptor (IL23R) ELISA Kit
Supplier:  Biomatik Corporation EKU05299
Encompass_Preferred
Rat Interleukin 23 Receptor (IL23R) ELISA Kit for detecting IL23R in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: F1LX96. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7307