Learn More
Biomatik Corporation Rat Interleukin 23 Subunit Alpha (IL23a) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11184
Encompass_Preferred
Rat Interleukin 23 Subunit Alpha (IL23a) Mini sample ELISA Kit for detecting IL23a in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.6pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q91Z84. Gene ID: 155140. Target Synonyms: P19; IL23P19; SGRF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8202