Learn More
Biomatik Corporation Rat Interleukin 25 (IL25) CLIA Kit
Supplier:  Biomatik Corporation EKU09693
Encompass_Preferred
Rat Interleukin 25 (IL25) CLIA Kit for detecting IL25 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.47pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: D3ZLB1. Gene ID: 501996. Target Synonyms: IL17E; IL17-E; Interleukin-17E. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8465