Learn More
American Research Products Inc Rat Interleukin 33 IL 33 ELI
Catalog No. 5000328689
Encompass
Rat Interleukin 33 (IL-33) ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28689 Supplier American Research Products Inc Supplier No. CSBE14077R
Rat Interleukin 33 (IL-33) ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich