Learn More
Biomatik Corporation Rat Interleukin 33 (IL33) CLIA Kit
Supplier:  Biomatik Corporation EKU09815
Encompass_Preferred
Rat Interleukin 33 (IL33) CLIA Kit for detecting IL33 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.54pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q66H70. Gene ID: 361749. Target Synonyms: DV27; C9ORF26; IL1F11; NFHEV; Interleukin-1 Family, Member 11; Nuclear factor from high endothelial venules. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8430