Learn More
American Research Products Inc Rat Interleukin 35 IL 35 ELI
Catalog No. 5000329425
Encompass
Rat Interleukin 35 (IL-35) ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 1 56 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29425 Supplier American Research Products Inc Supplier No. CSBE13652R
Rat Interleukin 35 (IL-35) ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 1 56 pg/mL Sample serum plasma tissue homogenates Method Sandwich