Learn More
American Research Products Inc Rat Interleukin 6 receptor IL
Catalog No. 5000329215
Encompass
Rat Interleukin 6 receptor IL-6R ELISA Kit from CUSABIO Detection Range 0 625 ng/mL-40 ng/mL Sensitivity 0 156 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29215 Supplier American Research Products Inc Supplier No. CSBE11876R
Rat Interleukin 6 receptor IL-6R ELISA Kit from CUSABIO Detection Range 0 625 ng/mL-40 ng/mL Sensitivity 0 156 ng/mL Sample serum plasma tissue homogenates Method Sandwich