Learn More
Biomatik Corporation Rat Interleukin 7 (IL7) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11206
Encompass_Preferred
Rat Interleukin 7 (IL7) Mini sample ELISA Kit for detecting IL7 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P56478. Target Synonyms: Pre-B-cell Growth Factor; Lymphopoietin-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7240