Learn More
Biomatik Corporation Rat Interleukin 7 Receptor (IL7R) ELISA Kit
Supplier:  Biomatik Corporation EKU05373
Encompass_Preferred
Rat Interleukin 7 Receptor (IL7R) ELISA Kit for detecting IL7R in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.116ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: D4A3X8. Gene ID: 294797. Target Synonyms: CDW127; CD127; IL-7R-Alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8909