Learn More
BIOHIPPO Rat IREB2 Microsample
SDP
Catalog No. 5000315474
Encompass
Rat IREB2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IREB2 in Tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 105 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat IREB2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat IREB2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat IREB2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15474 Supplier BIOHIPPO Supplier No. BHE15210985
Rat IREB2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IREB2 in Tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 105 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat IREB2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat IREB2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat IREB2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color