Learn More
Biomatik Corporation Rat Iron Responsive Element Binding Protein 2 (IREB2) ELISA Kit
Supplier:  Biomatik Corporation EKU05399
Encompass_Preferred
Rat Iron Responsive Element Binding Protein 2 (IREB2) ELISA Kit for detecting IREB2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.112ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q62751. Gene ID: 64831. Target Synonyms: IRP2; Iron regulatory protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8908