Learn More
Biomatik Corporation Rat Ischemia Modified Albumin (IMA) ELISA Kit
Supplier:  Biomatik Corporation EKU10145
Encompass_Preferred
Rat Ischemia Modified Albumin (IMA) ELISA Kit for detecting IMA in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 2.47-200ug/ml. Sensitivity: 0.92ug/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8366