Learn More
American Research Products Inc Rat Ischemia Modified Albumin
Catalog No. 5000328411
Encompass
Rat Ischemia Modified Albumin IMA ELISA Kit from CUSABIO Detection Range 0 9 U/mL-60 U/mL Sensitivity 0 225 U/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-28411 Supplier American Research Products Inc Supplier No. CSBE13620R
Rat Ischemia Modified Albumin IMA ELISA Kit from CUSABIO Detection Range 0 9 U/mL-60 U/mL Sensitivity 0 225 U/mL Sample serum plasma tissue homogenates Method Competitive