Learn More
Biomatik Corporation Rat Isocitrate Dehydrogenase 1, Soluble (IDH1) ELISA Kit
Supplier:  Biomatik Corporation EKU05407
Encompass_Preferred
Rat Isocitrate Dehydrogenase 1, Soluble (IDH1) ELISA Kit for detecting IDH1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.35ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P41562. Gene ID: 24479. Target Synonyms: PICD; IDP; Oxalosuccinate decarboxylase; NADP (+)-specific ICDH; Cytosolic NADP-isocitrate dehydrogenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4914