Learn More
Sino Biological Rat JAM2 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 80376R02H100
Encompass_Preferred
A DNA sequence encoding the rat JAM2 (Q3MHC0) (Met1-Asn236) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-0778