Learn More
SAI Infusion Technologies Rounded Tip Rat Jugular Vein Catheter - 30cm PU for Rats 150g+
SDP
Catalog No. NC0189785
Encompass
Rounded Tip Jugular Vein Catheter for Rats 150g or larger. 30cm Rounded Tip Vascuthane PU catheter with 3cm 3Fr tip and 2 suture beads to anchor the catheter in place. Sterile. Access with a 22G blunt needle such as the B22-50. Product is for laboratory animal research only.
Catalog No. NC0189785 Supplier SAI Infusion Technologies Supplier No. RJVR10
Rounded Tip Jugular Vein Catheter for Rats 150g or larger. 30cm Rounded Tip Vascuthane PU catheter with 3cm 3Fr tip and 2 suture beads to anchor the catheter in place. Sterile. Access with a 22G blunt needle such as the B22-50. Product is for laboratory animal research only.