Learn More
SAI Infusion Technologies Rat Jugular Vein Catheter - 18cm tapered PU for Rats 150g+
SDP
Catalog No. NC1393797
Encompass
Jugular Vein Catheter for Rats 150g or larger. 18cm Tapered PU catheter with 1.5cm 3Fr tip and 2 suture beads to anchor the catheter in place. Sterile. Access with a 22G blunt needle such as the B22-50. Product is for laboratory animal research only.
Catalog No. NC1393797 Supplier SAI Infusion Technologies Supplier No. RJV02
Jugular Vein Catheter for Rats 150g or larger. 18cm Tapered PU catheter with 1.5cm 3Fr tip and 2 suture beads to anchor the catheter in place. Sterile. Access with a 22G blunt needle such as the B22-50. Product is for laboratory animal research only.