Learn More
SAI Infusion Technologies Rat Jugular Vein Catheter - 17.8cm PU
SDP
Catalog No. NC1393798
Encompass
Jugular Vein Catheter for Rats. 17.8cm PU catheter with 2.8cm 3Fr tip and 2 suture beads to anchor the catheter in place. Sterile. Access with a 22G blunt needle such as the B22-50. Product is for laboratory animal research only.
Catalog No. NC1393798 Supplier SAI Infusion Technologies Supplier No. RJV04
Jugular Vein Catheter for Rats. 17.8cm PU catheter with 2.8cm 3Fr tip and 2 suture beads to anchor the catheter in place. Sterile. Access with a 22G blunt needle such as the B22-50. Product is for laboratory animal research only.