Learn More
Biomatik Corporation Rat Kallikrein 4 (KLK4) ELISA Kit
Supplier:  Biomatik Corporation EKU05442
Encompass_Preferred
Rat Kallikrein 4 (KLK4) ELISA Kit for detecting KLK4 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.049ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6IE12. Gene ID: 408210. Target Synonyms: ARM1; EMSP1; KLK-L1; PRSS17; PSTS; Enamel Matrix,Prostate; Kallikrein-Related Peptidase 4; Enamel matrix serine proteinase 1; Serine protease 17; Kallikrein-like protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7669