Learn More
Biomatik Corporation Rat Kallikrein 9 (KLK9) ELISA Kit
Supplier:  Biomatik Corporation EKU05454
Encompass_Preferred
Rat Kallikrein 9 (KLK9) ELISA Kit for detecting KLK9 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.052ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: G3V934. Gene ID: 292851. Target Synonyms: KLK-L3; Kallikrein-Related Peptidase 9; Kallikrein-like protein 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5982