Learn More
American Research Products Inc Rat keratinocyte autocrine fac
Catalog No. 5000326489
Encompass
Rat keratinocyte autocrine factor/Amphiregulin KAF/AR ELISA Kit from CUSABIO Detection Range 1 25 pg/mL-80 pg/mL Sensitivity 0 31 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26489 Supplier American Research Products Inc Supplier No. CSBE07299R
Rat keratinocyte autocrine factor/Amphiregulin KAF/AR ELISA Kit from CUSABIO Detection Range 1 25 pg/mL-80 pg/mL Sensitivity 0 31 pg/mL Sample serum plasma tissue homogenates Method Sandwich