Learn More
American Research Products Inc Rat Keratinocyte Growth Factor
Supplier:  American Research Products Inc CSBE12905R
Encompass
Rat Keratinocyte Growth Factor KGF ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-29947