Learn More
American Research Products Inc Rat Kidney injury molecule 1
Catalog No. 5000329224
Encompass
Rat Kidney injury molecule 1 Kim-1 ELISA Kit from CUSABIO Detection Range 0 313 ng/mL-20 ng/mL Sensitivity 0 228 ng/mL Sample serum plasma urine Method Sandwich
Catalog No. 50-003-29224 Supplier American Research Products Inc Supplier No. CSBE08808R
Rat Kidney injury molecule 1 Kim-1 ELISA Kit from CUSABIO Detection Range 0 313 ng/mL-20 ng/mL Sensitivity 0 228 ng/mL Sample serum plasma urine Method Sandwich