Learn More
Biomatik Corporation Rat Kidney Injury Molecule 1 (Kim1) CLIA Kit
Supplier:  Biomatik Corporation EKU09472
Encompass_Preferred
Rat Kidney Injury Molecule 1 (Kim1) CLIA Kit for detecting Kim1 in urine, tissue homogenates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 5.49-4,000pg/ml. Sensitivity: 2.33pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: O54947. Gene ID: 286934. Target Synonyms: HAVCR1; TIM1; TIMD1; HAVCR; Hepatitis A Virus Cellular Receptor 1; T Cell Immunoglobulin And Mucin Domain-Containing Protein 1; T-cell immunoglobulin mucin receptor 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5291