Learn More
Biomatik Corporation Rat Kinesin Family, Member 5B (KIF5B) ELISA Kit
Supplier:  Biomatik Corporation EKU05500
Encompass_Preferred
Rat Kinesin Family, Member 5B (KIF5B) ELISA Kit for detecting KIF5B in tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q2PQA9. Gene ID: 117550. Target Synonyms: KNS1; UKHC; KINH; Conventional kinesin heavy chain; Ubiquitous kinesin heavy chain; Kinesin-1 heavy chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5952