Learn More
BIOHIPPO Rat KMO Microsample
SDP
Catalog No. 5000313744
Encompass
Rat KMO Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of KMO in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 78-50 ng/mL with a sensitivity of 0 414 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat KMO Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat KMO Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat KMO biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13744 Supplier BIOHIPPO Supplier No. BHE15210826
Rat KMO Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of KMO in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 78-50 ng/mL with a sensitivity of 0 414 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat KMO Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat KMO Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat KMO biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color