Learn More
Biomatik Corporation Rat Kynurenine-3-Monooxygenase (KMO) ELISA Kit
Supplier:  Biomatik Corporation EKU08493
Encompass_Preferred
Rat Kynurenine-3-Monooxygenase (KMO) ELISA Kit for detecting KMO in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O88867. Gene ID: 59113. Target Synonyms: Kynurenine 3-Hydroxylase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7754