Learn More
Crystal Chem Inc Rat L-FABP ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80685
Encompass
The Rat L-FABP ELISA is a highly sensitive assay that is used to quantify levels of rat L-FABP in serum and plasma. Highlights: Kit uses sample size of only 20 µL. High sensitivity of 1.28 ng/mL. Dynamic range of12.5 - 400 ng/mL. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7812